C12H316CEE1E Chemcia Scientific, LLC

Pyrazolo[1,5-a]pyrimidin-6-ol

  • MDL number MFCD28096680
  • Cas Number : 1580489-59-3
  • Molecular Weight : 135.12625

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=4862

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4862
Availability details for
Quantity
  • Availability
  • Add to Cart