C12H316CEE21 Chemcia Scientific, LLC

3-Amino-4-methoxy-benzo[b]thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD08706234
  • Cas Number : 35212-88-5
  • Molecular Weight : 237.27922

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4865

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4865
Availability details for
Quantity
  • Availability
  • Add to Cart