C12H316CEE27 Chemcia Scientific, LLC

5-Bromo-3-iodo-2H-indazole

  • MDL number MFCD22123463
  • Cas Number : 459133-66-5
  • Molecular Weight : 322.93173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4873

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4873
Availability details for
Quantity
  • Availability
  • Add to Cart