C12H316CEE2B Chemcia Scientific, LLC

(4-Amino-3-iodo-phenyl)-acetonitrile

  • MDL number MFCD18389690
  • Cas Number : 955397-24-7
  • Molecular Weight : 258.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-amino-3-iodophenyl)acetonitrile
Inchi Code 1S/C8H7IN2/c9-7-5-6(3-4-10)1-2-8(7)11/h1-2,5H,3,11H2
Inchi Key LGPDBNNRUCQSBY-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5055

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5055
Availability details for
Quantity
  • Availability
  • Add to Cart