C12H316CEE2D Chemcia Scientific, LLC

6-Amino-3-iodo-5-nitro-1H-pyridin-2-one

  • MDL number MFCD11616172
  • Cas Number : 642460-96-6
  • Molecular Weight : 281.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-3-iodo-5-nitro-2(1H)-pyridinone
Inchi Code 1S/C5H4IN3O3/c6-2-1-3(9(11)12)4(7)8-5(2)10/h1H,(H3,7,8,10)
Inchi Key YPSCFDIDAMTMKR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5057

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5057
Availability details for
Quantity
  • Availability
  • Add to Cart