C12H316CEE31 Chemcia Scientific, LLC

6-Bromo-8-chloro-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine

  • MDL number MFCD28400727
  • Cas Number : 1649478-43-2
  • Molecular Weight : 247.48258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5061

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5061
Availability details for
Quantity
  • Availability
  • Add to Cart