C12H316CEE32 Chemcia Scientific, LLC

Acetic acid, cyano[[[(4-methylphenyl)sulfonyl]oxy]amino]-, ethyl ester

  • MDL number MFCD28400728
  • Cas Number : 91158-65-5
  • Molecular Weight : 298.31978

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5062

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5062
Availability details for
Quantity
  • Availability
  • Add to Cart