C12H316CEE33 Chemcia Scientific, LLC

5-Fluoro-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid ethyl ester

  • MDL number MFCD24387417
  • Cas Number : 185011-40-9
  • Molecular Weight : 235.2168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5063

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5063
Availability details for
Quantity
  • Availability
  • Add to Cart