C12H316CEE36 Chemcia Scientific, LLC

8-Chloro-imidazo[1,2-a]pyridine-2-carbaldehyde

  • Cas Number : 885276-03-9
  • Molecular Weight : 180.59485

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Green Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5009

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5009
Availability details for
Quantity
  • Availability
  • Add to Cart