C12H316CEE38 Chemcia Scientific, LLC

6-Amino-3-bromo-5-nitro-1H-pyridin-2-one

  • MDL number MFCD28400738
  • Cas Number : 1935346-20-5
  • Molecular Weight : 234.00993

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5065

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5065
Availability details for
Quantity
  • Availability
  • Add to Cart