C12H316CEE3A Chemcia Scientific, LLC

2-Amino-5-bromo-isonicotinonitrile

  • MDL number MFCD09907918
  • Cas Number : 944401-72-3
  • Molecular Weight : 198.02288

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5067

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5067
Availability details for
Quantity
  • Availability
  • Add to Cart