C12H316CEE40 Chemcia Scientific, LLC

5-Iodo-2H-pyrazolo[3,4-b]pyridin-3-ylamine

  • MDL number MFCD28049781
  • Cas Number : 1392152-87-2
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C6H5IN4/c7-3-1-4-5(8)10-11-6(4)9-2-3/h1-2H,(H3,8,9,10,11)
Inchi Key BYEUNDASYOFKIW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5073

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5073
Availability details for
Quantity
  • Availability
  • Add to Cart