C12H316CEE5A Chemcia Scientific, LLC

4-tert-Butyl-1-chloro-2-iodo-benzene

  • MDL number MFCD27980171
  • Cas Number : 1233062-20-8
  • Molecular Weight : 294.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(tert-butyl)-1-chloro-2-iodobenzene
Inchi Code 1S/C10H12ClI/c1-10(2,3)7-4-5-8(11)9(12)6-7/h4-6H,1-3H3
Inchi Key CFMXITGBZNSYTE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Colorless Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5077

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5077
Availability details for
Quantity
  • Availability
  • Add to Cart