C12H316CEE5B Chemcia Scientific, LLC

4-Amino-isothiazole-3,5-dicarboxylic acid 3-ethyl ester 5-methyl ester

  • MDL number MFCD03990496
  • Cas Number : 632291-73-7
  • Molecular Weight : 230.2439

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5078

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5078
Availability details for
Quantity
  • Availability
  • Add to Cart