C12H316CEE5D Chemcia Scientific, LLC

6-Methyl-3H-benzooxazole-2-thione

  • MDL number MFCD00175644
  • Cas Number : 23417-29-0
  • Molecular Weight : 165.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methyl-1,3-benzoxazole-2-thiol
Inchi Code 1S/C8H7NOS/c1-5-2-3-6-7(4-5)10-8(11)9-6/h2-4H,1H3,(H,9,11)
Inchi Key NUMDWMDMUXZMHA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5080

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5080
Availability details for
Quantity
  • Availability
  • Add to Cart