C12H316CEE62 Chemcia Scientific, LLC

5-Methyl-2H-indazol-3-ylamine

  • MDL number MFCD28400754
  • Cas Number : 88805-94-1
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-2H-indazol-3-amine
Inchi Code 1S/C8H9N3/c1-5-2-3-7-6(4-5)8(9)11-10-7/h2-4H,1H3,(H3,9,10,11)
Inchi Key UKKWIOCVCZXOAM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5048

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5048
Availability details for
Quantity
  • Availability
  • Add to Cart