C12H316CEE65 Chemcia Scientific, LLC

1-Iodo-3-methoxy-2-nitro-benzene

  • MDL number MFCD18397933
  • Cas Number : 725266-66-0
  • Molecular Weight : 279.03517

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5035

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5035
Availability details for
Quantity
  • Availability
  • Add to Cart