C12H316CEE66 Chemcia Scientific, LLC

5-Chloro-2-methyl-1H-imidazo[4,5-b]pyridine

  • MDL number MFCD02178641
  • Cas Number : 40851-92-1
  • Molecular Weight : 167.59897

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5049

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5049
Availability details for
Quantity
  • Availability
  • Add to Cart