C12H316CEE68 Chemcia Scientific, LLC

6-Iodo-5-methyl-imidazo[1,2-a]pyridine

  • MDL number MFCD28099867
  • Molecular Weight : 258.06279

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5038

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5038
Availability details for
Quantity
  • Availability
  • Add to Cart