C12H316CEE6B Chemcia Scientific, LLC

(S)-2-Isocyanomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD28967990
  • Cas Number : 1245908-97-7
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (S)-2-(isocyanomethyl)pyrrolidine-1-carboxylate
Inchi Code 1S/C11H18N2O2/c1-11(2,3)15-10(14)13-7-5-6-9(13)8-12-4/h9H,5-8H2,1-3H3/t9-/m0/s1
Inchi Key RHJWPRSOYYDGJX-VIFPVBQESA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Colorless Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5128

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5128
Availability details for
Quantity
  • Availability
  • Add to Cart