C12H316CEE6C Chemcia Scientific, LLC

2,6-Dimethyl-1-oxy-pyridine-3,5-dicarboxylic acid diethyl ester

  • MDL number MFCD28400763
  • Cas Number : 33521-04-9
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-bis(ethoxycarbonyl)-2,6-dimethylpyridine 1-oxide
Inchi Code 1S/C13H17NO5/c1-5-18-12(15)10-7-11(13(16)19-6-2)9(4)14(17)8(10)3/h7H,5-6H2,1-4H3
Inchi Key QUHNZPKYBBTNFQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5051

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5051
Availability details for
Quantity
  • Availability
  • Add to Cart