C12H316CEE71 Chemcia Scientific, LLC

1-(2-Chloro-5-methoxy-phenyl)-ethanone

  • MDL number MFCD14534275
  • Cas Number : 77344-69-5
  • Molecular Weight : 184.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-chloro-5-methoxyphenyl)ethanone
Inchi Code 1S/C9H9ClO2/c1-6(11)8-5-7(12-2)3-4-9(8)10/h3-5H,1-2H3
Inchi Key WORVKDABKDAROB-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5087

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5087
Availability details for
Quantity
  • Availability
  • Add to Cart