C12H316CEE7A Chemcia Scientific, LLC

2-Amino-5-thiocyanato-benzoic acid methyl ester

  • MDL number MFCD03176138
  • Cas Number : 117324-16-0
  • Molecular Weight : 208.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-5-thiocyanatobenzoate
Inchi Code 1S/C9H8N2O2S/c1-13-9(12)7-4-6(14-5-10)2-3-8(7)11/h2-4H,11H2,1H3
Inchi Key RGPZBRDJSOXWJT-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5093

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5093
Availability details for
Quantity
  • Availability
  • Add to Cart