C12H316CEE7D Chemcia Scientific, LLC

(6-Iodo-pyridin-3-yl)-dimethyl-amine

  • Molecular Weight : 248.06758

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5097

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5097
Availability details for
Quantity
  • Availability
  • Add to Cart