C12H316CEE7E Chemcia Scientific, LLC

3-Methyl-5-nitro-benzo[d]isoxazole

  • Cas Number : 63770-48-9
  • Molecular Weight : 178.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methyl-5-nitro-1,2-benzisoxazole
Inchi Code 1S/C8H6N2O3/c1-5-7-4-6(10(11)12)2-3-8(7)13-9-5/h2-4H,1H3
Inchi Key PQLIRZDJBWDLOU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5100

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5100
Availability details for
Quantity
  • Availability
  • Add to Cart