C12H316CEE7F Chemcia Scientific, LLC

2-Chloromethyl-5-fluoro-pyridine

  • MDL number MFCD10698608
  • Cas Number : 915690-76-5
  • Molecular Weight : 145.56485

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5101

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5101
Availability details for
Quantity
  • Availability
  • Add to Cart