C12H316CEE82 Chemcia Scientific, LLC

1-(2-Methoxy-5-nitro-phenyl)-ethanone

  • Cas Number : 61941-46-6
  • Molecular Weight : 195.17638

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5110

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5110
Availability details for
Quantity
  • Availability
  • Add to Cart