C12H316CEE86 Chemcia Scientific, LLC

3-Methyl-1-oxy-pyridine-2-carbonitrile

  • Cas Number : 159727-88-5
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methyl-2-pyridinecarbonitrile 1-oxide
Inchi Code 1S/C7H6N2O/c1-6-3-2-4-9(10)7(6)5-8/h2-4H,1H3
Inchi Key OTXYVTPSEMLVLS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5118

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5118
Availability details for
Quantity
  • Availability
  • Add to Cart