C12H316CEE87 Chemcia Scientific, LLC

4-Methyl-1H-indazol-3-ylamine

  • MDL number MFCD19215920
  • Cas Number : 1207529-96-1
  • Molecular Weight : 147.18103

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5123

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5123
Availability details for
Quantity
  • Availability
  • Add to Cart