C12H316CEE8D Chemcia Scientific, LLC

7-Methyl-3,4-dihydro-2H-benzo[1,4]oxazine

  • MDL number MFCD17014030
  • Cas Number : 71472-58-7
  • Molecular Weight : 149.19412

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5137

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5137
Availability details for
Quantity
  • Availability
  • Add to Cart