C12H316CEE8E Chemcia Scientific, LLC

1-Iodo-4-nitro-2-trifluoromethyl-benzene

  • MDL number MFCD09800734
  • Cas Number : 400-75-9
  • Molecular Weight : 317.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-iodo-4-nitro-2-(trifluoromethyl)benzene
Inchi Code 1S/C7H3F3INO2/c8-7(9,10)5-3-4(12(13)14)1-2-6(5)11/h1-3H
Inchi Key HUFFQIQKEINJNA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5138

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5138
Availability details for
Quantity
  • Availability
  • Add to Cart