C12H316CEE8F Chemcia Scientific, LLC

3-Amino-5-chloro-benzo[b]thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD13689054
  • Cas Number : 1026241-99-5
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-amino-5-chlorobenzo[b]thiophene-2-carboxylate
Inchi Code 1S/C10H8ClNO2S/c1-14-10(13)9-8(12)6-4-5(11)2-3-7(6)15-9/h2-4H,12H2,1H3
Inchi Key CFYACBCHHBWDOO-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5139

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5139
Availability details for
Quantity
  • Availability
  • Add to Cart