C12H316CEE91 Chemcia Scientific, LLC

3-(2-tert-Butoxycarbonylamino-ethylcarbamoyl)-acrylic acid

  • MDL number MFCD28968002
  • Cas Number : 215654-55-0
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (Z)-4-((2-((tert-butoxycarbonyl)amino)ethyl)amino)-4-oxobut-2-enoic acid
Inchi Code 1S/C11H18N2O5/c1-11(2,3)18-10(17)13-7-6-12-8(14)4-5-9(15)16/h4-5H,6-7H2,1-3H3,(H,12,14)(H,13,17)(H,15,16)/b5-4-
Inchi Key INGPNYUYNRFJBP-PLNGDYQASA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin CN
Product Link http://www.chemcia.com/product_fl02.asp?id=5141

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5141
Availability details for
Quantity
  • Availability
  • Add to Cart