C12H316CEE95 Chemcia Scientific, LLC

2-Bromo-4-nitro-6-trifluoromethyl-phenylamine

  • MDL number MFCD14560537
  • Cas Number : 400-66-8
  • Molecular Weight : 285.02133

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5144

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5144
Availability details for
Quantity
  • Availability
  • Add to Cart