C12H316CEE97 Chemcia Scientific, LLC

(5-Chloro-3-nitro-pyridin-2-yl)-hydrazine

  • MDL number MFCD10000082
  • Cas Number : 22353-46-4
  • Molecular Weight : 188.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-hydrazino-3-nitropyridine
Inchi Code 1S/C5H5ClN4O2/c6-3-1-4(10(11)12)5(9-7)8-2-3/h1-2H,7H2,(H,8,9)
Inchi Key UHYSNWKUKBMEHR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Reddish-Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5146

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5146
Availability details for
Quantity
  • Availability
  • Add to Cart