C12H316CEE9D Chemcia Scientific, LLC

N'-Cyclopentylidene-hydrazinecarboxylic acid tert-butyl ester

  • MDL number MFCD00471042
  • Cas Number : 79201-39-1
  • Molecular Weight : 198.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-cyclopentylidenehydrazinecarboxylate
Inchi Code 1S/C10H18N2O2/c1-10(2,3)14-9(13)12-11-8-6-4-5-7-8/h6,11H,4-5,7H2,1-3H3,(H,12,13)
Inchi Key AOMYVPFREKTMOP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin CN
Product Link http://www.chemcia.com/product_fl02.asp?id=5152

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5152
Availability details for
Quantity
  • Availability
  • Add to Cart