C12H316CEEA6 Chemcia Scientific, LLC

6-Amino-5-methoxy-pyridine-2-carboxylic acid methyl ester

  • MDL number MFCD18261109
  • Cas Number : 1072077-54-3
  • Molecular Weight : 182.1805

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5172

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5172
Availability details for
Quantity
  • Availability
  • Add to Cart