C12H316CEEAE Chemcia Scientific, LLC

7-Chloro-1H-pyrido[2,3-b][1,4]oxazin-2-one

  • MDL number MFCD01648626
  • Cas Number : 105544-33-0
  • Molecular Weight : 184.5831

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5250

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5250
Availability details for
Quantity
  • Availability
  • Add to Cart