C12H316CEEB1 Chemcia Scientific, LLC

2-Hydroxy-6-methyl-nicotinic acid methyl ester

  • MDL number MFCD09953605
  • Cas Number : 51146-06-6
  • Molecular Weight : 167.16583

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5255

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5255
Availability details for
Quantity
  • Availability
  • Add to Cart