C12H316CEEB3 Chemcia Scientific, LLC

4-Oxo-1,4-dihydro-quinoline-8-carbonitrile

  • MDL number MFCD26131872
  • Cas Number : 127285-55-6
  • Molecular Weight : 170.17212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5260

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5260
Availability details for
Quantity
  • Availability
  • Add to Cart