C12H316CEEB5 Chemcia Scientific, LLC

3-Iodo-4-oxo-1,4-dihydro-quinoline-6-carboxylic acid

  • Molecular Weight : 315.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydroxy-3-iodoquinoline-6-carboxylic acid
Inchi Code 1S/C10H6INO3/c11-7-4-12-8-2-1-5(10(14)15)3-6(8)9(7)13/h1-4H,(H,12,13)(H,14,15)
Inchi Key JSSDYRDNIWICTD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5217

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5217
Availability details for
Quantity
  • Availability
  • Add to Cart