C12H316CEEBA Chemcia Scientific, LLC

5-Methoxy-2H-indazol-3-ylamine

  • MDL number MFCD28539846
  • Molecular Weight : 163.18043

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5222

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5222
Availability details for
Quantity
  • Availability
  • Add to Cart