C12H316CEEC5 Chemcia Scientific, LLC

(4-Bromo-5-fluoro-2-nitro-phenyl)-methanol

  • MDL number MFCD28740129
  • Cas Number : 1132682-66-6
  • Molecular Weight : 250.0252

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5245

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5245
Availability details for
Quantity
  • Availability
  • Add to Cart