C12H316CEECC Chemcia Scientific, LLC

4-Iodo-3-trifluoromethyl-phenylamine

  • MDL number MFCD04972659
  • Cas Number : 155403-06-8
  • Molecular Weight : 287.0242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5262

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5262
Availability details for
Quantity
  • Availability
  • Add to Cart