C12H316CEECD Chemcia Scientific, LLC

4-Chloro-2-dimethylaminomethylene-3-oxo-butyric acid ethyl ester

  • MDL number MFCD28968014
  • Cas Number : 1246616-68-1
  • Molecular Weight : 219.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (Z)-4-chloro-2-((dimethylamino)methylene)-3-oxobutanoate
Inchi Code 1S/C9H14ClNO3/c1-4-14-9(13)7(6-11(2)3)8(12)5-10/h6H,4-5H2,1-3H3/b7-6-
Inchi Key DMPAUMVQAGFPRL-SREVYHEPSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Black Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5257

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5257
Availability details for
Quantity
  • Availability
  • Add to Cart