C12H316CEECE Chemcia Scientific, LLC

3-Methoxy-4-methyl-2,6-dinitro-phenol

  • MDL number MFCD28968015
  • Cas Number : 1951440-98-4
  • Molecular Weight : 228.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methoxy-4-methyl-2,6-dinitrophenol
Inchi Code 1S/C8H8N2O6/c1-4-3-5(9(12)13)7(11)6(10(14)15)8(4)16-2/h3,11H,1-2H3
Inchi Key ZUZCDRVWQHPLOJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5258

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5258
Availability details for
Quantity
  • Availability
  • Add to Cart