C12H316CEECF Chemcia Scientific, LLC

3-(4-Methoxy-benzyloxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzonitrile

  • MDL number MFCD28968016
  • Molecular Weight : 365.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-((4-methoxybenzyl)oxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Inchi Code 1S/C21H24BNO4/c1-20(2)21(3,4)27-22(26-20)17-10-16(13-23)11-19(12-17)25-14-15-6-8-18(24-5)9-7-15/h6-12H,14H2,1-5H3
Inchi Key PLCZAJGZPMTJDL-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5249

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5249
Availability details for
Quantity
  • Availability
  • Add to Cart