C12H316CEED6 Chemcia Scientific, LLC

7-Chloro-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine

  • MDL number MFCD18250280
  • Cas Number : 1260667-70-6
  • Molecular Weight : 170.59964

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5276

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5276
Availability details for
Quantity
  • Availability
  • Add to Cart