C12H316CEED9 Chemcia Scientific, LLC

4-Bromo-2-iodo-1-methoxy-benzene

  • MDL number MFCD13186707
  • Cas Number : 98273-59-7
  • Molecular Weight : 312.93367

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5285

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5285
Availability details for
Quantity
  • Availability
  • Add to Cart