C12H316CEEDD Chemcia Scientific, LLC

3-Methoxy-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzonitrile

  • MDL number MFCD11855976
  • Cas Number : 1035266-33-1
  • Molecular Weight : 259.11546

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5293

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5293
Availability details for
Quantity
  • Availability
  • Add to Cart